missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEDGIN6, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
LEDGIN6 (CX05168) is a quinoline-based protein-protein interaction inhibitor of LEDGF/p75 and HIV integrase.
Specifications
Specifications
| Chemical Name or Material | LEDGIN6 |
| CAS | 957890-42-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20ClNO2 |
| Quantity | 50 mg |
| Synonym | CX05168 CX04328 |
| Solubility Information | DMSO : ≥ 100 mg/mL (282.61 mM) |
| SMILES | ClC1=CC=C(N=C(C)C(C(CCC)C(O)=O)=C2C3=CC=CC=C3)C2=C1 |
| Molecular Weight (g/mol) | 353.84 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction