missing translation for 'onlineSavingsMsg'
Learn More

LEDGF Antibody, Biotin, Novus Biologicals™

Product Code. 18528513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18528513 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18528513 Supplier Novus Biologicals Supplier No. NB11040560B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LEDGF Polyclonal antibody specifically detects LEDGF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEDGF
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CLL-associated antigen KW-7, Dense fine speckles 70 kDa protein, DFS70, LEDGFDFS 70, Lens epithelium-derived growth factor, MGC74712, p52, p75, PAIP, PC4 and SFRS1 interacting protein 1, PC4 and SFRS1 interacting protein 2, PC4 and SFRS1-interacting protein, PSIP2, transcriptional coactivator p52/p75, Transcriptional coactivator p75/p52
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human Lens Epithelium Derived Growth Factor/Transcriptional Coactivator p75 using the numbering given in entry NP_150091.2 (GeneID 11168).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11168
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.