missing translation for 'onlineSavingsMsg'
Learn More

LEDGF Antibody (762815) [DyLight 488], Novus Biologicals™

Product Code. 30736269 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736269 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736269 Supplier Novus Biologicals Supplier No. FAB3468K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LEDGF Monoclonal antibody specifically detects LEDGF in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEDGF
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 762815
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CLL-associated antigen KW-7, Dense fine speckles 70 kDa protein, DFS70, LEDGFDFS 70, Lens epithelium-derived growth factor, MGC74712, p52, p75, PAIP, PC4 and SFRS1 interacting protein 1, PC4 and SFRS1 interacting protein 2, PC4 and SFRS1-interacting protein, PSIP2, transcriptional coactivator p52/p75, Transcriptional coactivator p75/p52
Host Species Mouse
Immunogen E. coli-derived recombinant human LEDGF, Met1-Lys333, Accession # O75475
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11168
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.