missing translation for 'onlineSavingsMsg'
Learn More

LED209, MedChemExpress

Product Code. 16493827
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493827 50 mg 50mg
16417888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16493827 Supplier Medchem Express Supplier No. HY19748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LED209 is a potent small molecule inhibitor of bacterial receptor QseC, is a potent prodrug that is highly selective for QseC.

LED209 is a potent small molecule inhibitor of bacterial receptor QseC, is a potent prodrug that is highly selective for QseC.

Specifications

Chemical Name or Material LED209
CAS 245342-14-7
Color White
Purity Grade Notes Research
Molecular Formula C19H17N3O2S2
Quantity 50 mg
Solubility Information H2O : 1 mg/mL (2.61 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (2.61 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMILES O=S(C1=CC=C(NC(NC2=CC=CC=C2)=S)C=C1)(NC3=CC=CC=C3)=O
Molecular Weight (g/mol) 383.49
Formula Weight 383.49
Percent Purity 95.66%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.