missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ LED Module for Syngene Gbox Chemi System
GREENER_CHOICE

Product Code. 15829962 Shop All Syngene Products
Click to view available options
Wavelength:
465nm
520nm
635nm
740nm
Color:
Blue
Green
Infra Red
Red
Product Type:
LED Module
LED Module, Multiplex
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15829962

Brand: Syngene™ LEDGREEN x 2

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Syngene™ LED Module for Syngene Gbox Chemi System for western fluorescence

Specifications

Color Green
For Use With (Equipment) Gbox Chemi Systems
Quantity 2
Wavelength 520nm
Product Type LED Module
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.