Learn More
Motic™ LED Module for Epi Illuminator
Click to view available options
Description:
6000°K
Unit Size:
Each
Description
- Long-lasting and powerful LED.
- Available in two color temperatures (4500°K, and 6000°K).
Specifications
Specifications
| Type | LED Module |
| Description | 6000°K |
| For Use With (Equipment) | Epi Illuminator, BA410E, BA310MET and AE31E microscopes |
| Voltage | 12 V |