Learn More
Motic™ LED Module, BA210
Click to view available options
For Use With (Equipment):
BA210 Series Upright Compound Microscopes
Unit Size:
Each
Description
- BA210 Microscope lighting module.
- Long-lasting LED.
- 6000°K color temperature
Specifications
Specifications
| Type | LED Transmitted Light Module |
| Description | 6000°K ±300°K |
| For Use With (Equipment) | BA210 Series Upright Compound Microscopes |
| Voltage | 6 V |