missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek LED Cubes for Imaging Multi-Mode Reader

Product Code. 15391480 Shop All Agilent BioTek Products
Click to view available options
Light Source:
390 nm LED cube
655 nm LED cube
CY5 LED
CY7 LED
GFP/CFP LED
RFP LED
Texas Red LED
YFP LED
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15391480

Brand: Agilent BioTek 1225001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Imaging Multi-Mode Readers.

For use with Imaging Multi-Mode Readers.
TRUSTED_SUSTAINABILITY

Specifications

Type LED Light Cube
For Use With (Equipment) Imaging Multi-Mode Reader
Color Green/Cyan
Light Source GFP/CFP LED
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.