missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LECT2 Antibody (MM0449-4B24), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11768H
This item is not returnable.
View return policy
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LECT2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| LECT2 | |
| Human recombinant LECT2 | |
| 0.1 mL | |
| 3950 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| MM0449-4B24 | |
| Western Blot, Immunohistochemistry | |
| chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction