missing translation for 'onlineSavingsMsg'
Learn More

LECT2 Antibody (MM0449-4B24), Allophycocyanin, Novus Biologicals™

Product Code. 18403422 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403422 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403422 Supplier Novus Biologicals Supplier No. NBP211768APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LECT2 Monoclonal antibody specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LECT2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone MM0449-4B24
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628
Host Species Mouse
Immunogen Human recombinant LECT2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3950
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.