missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MP Biomedicals™

Product Code. p-4679606 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 11388999

Brand: MP Biomedicals 0210214780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Molecular biology, Cell biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8030-76-0
Molecular Formula C11H22NO8P
Synonym L-a-Phosphatidylcholine

Specifications

Color Yellow
Quantity 100 g
Formula Weight 327.27
Physical Form Solid
Chemical Name or Material Lecithin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.