Learn More
Invitrogen™ Lebrikizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541906
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Lebrikizumab has been used in trials studying the treatment of Asthma, Allergic Asthma, Atopic Dermatitis, Idiopathic Pulmonary Fibrosis, and COPD, Chronic Obstructive Pulmonary Disease.
Specifications
| Lebrikizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MILR1444A; PRO301444 | |
| Human IL13. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG4 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.