missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Honeywell™ Electrosoft Latex Gloves

Product Code. 12697583
Change view
Click to view available options
For Use With (Application):
Maximum 26,500 volts
Maximum 500 volts
Size:
7
8
9
10
Product Type:
Class 00 Gloves
Class 3 Gloves
Thickness (Metric):
0.5 mm
2.9 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Size Product Type Thickness (Metric) unitSize
12697583 Maximum 26,500 volts 10 Class 3 Gloves 2.9 mm Each
12667553 Maximum 500 volts 10 Class 00 Gloves 0.5 mm Each
12687553 Maximum 500 volts 7 Class 00 Gloves 0.5 mm Each
12697553 Maximum 500 volts 8 Class 00 Gloves 0.5 mm Each
12607563 Maximum 500 volts 9 Class 00 Gloves 0.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12697583 Supplier Sperian™ Supplier No. 209193110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dielectric latex gloves to protect against electrical risks from 500 to 3600 volts, depending on model. Sperian™ Honeywell™ Electrosoft Latex Gloves have an ergonomic shape and lightly powdered interior to make gloves easy to slip on and off.

  • Rolled or cut edges
  • Needs leather overglove for mechanical protection
  • Natural latex base offers high dielectric characteristics
  • Recommneded for the transportation, utilities and wind energy industries
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Product Type Class 3 Gloves
Size 10
Thickness (Metric) 2.9 mm
For Use With (Application) Maximum 26,500 volts
Quantity 1 Each
Length (Metric) 36 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.