missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121800
This item is not returnable.
View return policy
Description
LEAP2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| LEAP2 | |
| Polyclonal | |
| Unconjugated | |
| LEAP2 | |
| LEAP2; LEAP-2; liver enriched antimicrobial peptide 2; liver expressed antimicrobial peptide 2; liver-expressed antimicrobial peptide 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116842, 259301, 396687, 497901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q91V13, Q95JB4, Q969E1 | |
| LEAP2 | |
| Recombinant protein Liver Expressed Antimicrobial Peptide 2. The antigen corresponds to amino acid range 23-76 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction