missing translation for 'onlineSavingsMsg'
Learn More

Lead zirconium titanium oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.9% (metals basis), Thermo Scientific™

Product Code. 15484835
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484835 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15484835 Supplier Thermo Scientific Alfa Aesar Supplier No. 041068.KS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Magnetron sputtering source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12626-81-2
Color White
Quantity 1 Ea.
Molecular Formula O5PbTiZr
MDL Number MFCD00212270
Synonym lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
InChI Key HFGPZNIAWCZYJU-UHFFFAOYSA-N
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]
IUPAC Name lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
Molecular Weight (g/mol) 426.286
PubChem CID 159452
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lead zirconium titanium oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead zirconium titanium oxide sputtering target
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.