missing translation for 'onlineSavingsMsg'
Learn More

Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals

Product Code. 11334737
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334737 10 g 10g
11344737 50 g 50g
11354737 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334737 Supplier Thermo Scientific Alfa Aesar Supplier No. 039758.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in piezoelectric devices and electro-optic devices

Lead zirconium titanium oxide is used in piezoelectric devices and electro-optic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12626-81-2
Molecular Formula O5PbTiZr
Molecular Weight (g/mol) 426.286
MDL Number MFCD00212270
InChI Key HFGPZNIAWCZYJU-UHFFFAOYSA-N
Synonym lead zirconate titanate, lead titanate zirconate, lead titanium zirconium oxide, lead zirconium titanate, pzt-c, plumbum zirconate-titanate, pzt 5, pzt 8, lead titanium zirconium oxide pb ti,zr o3, lead zirconium titanium oxide
PubChem CID 159452
IUPAC Name lead(2+);oxygen(2-);titanium(4+);zirconium(4+)
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Ti+4].[Zr+4].[Pb+2]

Specifications

Color Yellow
Physical Form Semi-solid, Viscous Liquid
Quantity 10 g
UN Number UN1993
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈30 wt%
Chemical Name or Material Lead zirconium titanium oxide, Polymeric precursor
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead zirconium titanium oxide, polymeric precursor
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Lead zirconium titanium oxide, Toluene,2-Propanol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.