missing translation for 'onlineSavingsMsg'
Learn More

Lead Titanium Zirconium Oxide (~30% by weight), TRC

Product Code. 30471678
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471678 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471678 Supplier Toronto Research Chemicals Supplier No. TRCL32001510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lead Titanium Zirconium Oxide
CAS 12626-81-2
Quantity 10 g
Molecular Formula O3 Pb Ti(x) Zr(y)
Synonym 3203HD, 5A4E, APC 840, APC 855, APC 856, BZ27M5, EBL 3, EPC 21, Fuji C 22, Fuji C 6, Lead Titanate Zirconate, Lead Titanate Zirconate (Pb(Ti,Zr)O3), Lead Titanate Zirconate (PbTi0-1Zr0-1O3), Lead Titanium Zirconium Oxide (PbTi0-1Zr0-1O3), Lead Zirconate Titanate, Motorola 3203HD, P 1-60, P 41, P 5, P 5 (oxide), PCR-7M, PE 600, PE 60A, PE 60E, PK 51, PKI 552, PZT, PZT 3203HD, PZT 5, PZT 501A, PZT 502, PZT 574, PZT 5A4, PZT 5H, PZT 8, PZT 81005A, PZT 850
InChI Formula InChI=1S/5O.Pb.Ti.Zr/q5*-2;+2;2*+4
SMILES OC(=O)CCCCCCC(=O)OC
Molecular Weight (g/mol) 394.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.