missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Titanium Zirconium Oxide (~30% by weight), TRC
Specifications
Specifications
| Chemical Name or Material | Lead Titanium Zirconium Oxide |
| CAS | 12626-81-2 |
| Quantity | 1 g |
| Molecular Formula | O3 Pb Ti(x) Zr(y) |
| Synonym | 3203HD, 5A4E, APC 840, APC 855, APC 856, BZ27M5, EBL 3, EPC 21, Fuji C 22, Fuji C 6, Lead Titanate Zirconate, Lead Titanate Zirconate (Pb(Ti,Zr)O3), Lead Titanate Zirconate (PbTi0-1Zr0-1O3), Lead Titanium Zirconium Oxide (PbTi0-1Zr0-1O3), Lead Zirconate Titanate, Motorola 3203HD, P 1-60, P 41, P 5, P 5 (oxide), PCR-7M, PE 600, PE 60A, PE 60E, PK 51, PKI 552, PZT, PZT 3203HD, PZT 5, PZT 501A, PZT 502, PZT 574, PZT 5A4, PZT 5H, PZT 8, PZT 81005A, PZT 850 |
| InChI Formula | InChI=1S/5O.Pb.Ti.Zr/q5*-2;+2;2*+4 |
| SMILES | OC(=O)CCCCCCC(=O)OC |
| Molecular Weight (g/mol) | 394.29 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?