missing translation for 'onlineSavingsMsg'
Learn More

Lead Tin, solder alloy, 3.18mm (0.125in) dia

Product Code. 11369427
Click to view available options
Quantity:
113 g
225 g
Unit Size:
113g
225g
This item is not returnable. View return policy

Product Code. 11369427

Brand: Thermo Scientific Alfa Aesar 041023.29

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Pb:Sn; 50:50 wt%

For use with copper, copper alloys, lead, steel, and nickel alloys. Recommended for light electrical wiring

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Pb:Sn; 50:50 wt%
MDL Number MFCD00798549

Specifications

Packaging Plastic bottle
Quantity 225 g
Chemical Name or Material Lead Tin
compliance-icons compliance-icons
Product Identifier
  • Lead Tin, solder alloy
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H362-May cause harm to breast-fed children.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.