Learn More
Lead Tin, solder alloy, 1.6mm (0.06in) dia, with rosin core, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041024.TB
Code : NEW
Additional Details : Weight : 0.11300kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead Tin, Solder Alloy | |
Pb:Sn; 50:50 wt% | |
113 g |
1.6mm (0.06 in.) dia., with rosin core | |
MFCD00798549 | |
Solid |
RUO – Research Use Only