missing translation for 'onlineSavingsMsg'
Learn More

Lead Tetraacetate, purum p.a., ≥95% (dried material), Honeywell Burdick & Jackson™

Product Code. p-7120506 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
15661450 100 g Glass Bottle 100g
15671450 500 g Glass Bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15661450

Brand: Honeywell Research Chemicals 15370100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a., moistened with ∼5% acetic acid to stabilize,≥95% (dried material)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-67-8
Molecular Formula C8H16O8Pb
Molecular Weight (g/mol) 447.408
MDL Number MFCD00008693
InChI Key NVTAREBLATURGT-UHFFFAOYSA-N
Synonym lead iv acetate
PubChem CID 50931538
IUPAC Name acetic acid;lead
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]

Specifications

Melting Point 180° to 190°
Color White
Quantity 100 g
Linear Formula Pb(CH3COO)4
UN Number UN2923
Beilstein 3595640
Formula Weight 443.38g/mol
Percent Purity ≥95% (dried material)
Grade purum p.a.
Packaging Glass Bottle
Appearance Crystals
Density 2.230g/cu.cm
Chemical Name or Material Lead Tetraacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.