missing translation for 'onlineSavingsMsg'
Learn More

Lead rod, 6.35mm (0.25in) dia, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11325437
Click to view available options
Quantity:
50 mm
250 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7439-92-1
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
5352425
CHEBI:27889
lead
[Pb]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7439-92-1
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
5352425
CHEBI:27889
lead
[Pb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Quantity 50 mm
Percent Purity 99.999%
Odor Odorless
Weight ≈18g/50mm
Form Rod
Assay (metals basis)
Chemical Name or Material Lead rod, 6.35mm (0.25 in.) dia.
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead rod
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'