missing translation for 'onlineSavingsMsg'
Learn More

Lead, Pure, Metal, Granulated, Fisher Chemical

Product Code. 10204300 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204300 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204300 Supplier Fisher Chemical Supplier No. L/0700/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7439-92-1
Melting Point 327.4°C
Boiling Point 1740°C
Quantity 500 g
Packaging Plastic powder jar
Form Solid
Molecular Formula Pb
MDL Number MFCD00134050
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES [Pb]
IUPAC Name lead
Molecular Weight (g/mol) 207.20
PubChem CID 5352425
ChEBI CHEBI:27889
Formula Weight 207.19g/mol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.