missing translation for 'onlineSavingsMsg'
Learn More

Lead Nitrate Solution, Reagecon™

Product Code. 11805782 Shop All Reagecon Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11805782

Brand: Reagecon PB2011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.1M
compliance-icons
Product Identifier
  • LEAD NITRATE 0.1M (0.2N)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : lead dinitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.