missing translation for 'onlineSavingsMsg'
Learn More

Lead molybdenum oxide, 99%, Thermo Scientific™

Product Code. 11474280
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474280 25 g 25g
11484280 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474280 Supplier Thermo Scientific Alfa Aesar Supplier No. A10576.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10190-55-3
Molecular Formula MoO4Pb
Molecular Weight (g/mol) 367.146
MDL Number MFCD00016272
InChI Key XJUNRGGMKUAPAP-UHFFFAOYSA-N
Synonym lead molybdate, unii-wgp849k028, lead molybdate vi, lead molybdenum oxide pbmoo4, moo4.pb, $l^ 2-lead 2+ ion molybdate, dioxido dioxo molybdenum; lead 2+, lead ii molybdate trace metals basis
PubChem CID 61490
IUPAC Name dioxido(dioxo)molybdenum;lead(2+)
SMILES [O-][Mo](=O)(=O)[O-].[Pb+2]

Specifications

Quantity 25 g
Linear Formula PbMoO4
UN Number UN3077
Formula Weight 367.13
Percent Purity 99%
Odor Odorless
Density 6.7
Chemical Name or Material Lead molybdenum oxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead molybdenum oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.