missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead lanthanum zirconium titanium oxide, polymeric precursor, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Lead lanthanum zirconium titanium oxide |
| Name Note | Polymeric precursor |
| Color | Brown |
| Physical Form | Viscous Liquid |
| Quantity | 1 g |
| MDL Number | MFCD02684260 |
| UN Number | UN1993 |
| Solubility Information | Soluble in hexane, toluene and alcohol |
| Sensitivity | Moisture sensitive |
Safety and Handling
Product Identifier
- Lead lanthanum zirconium titanium oxide, polymeric precursor
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?