missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) acetate, 95%, stabilized

Product Code. p-4509518 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10644872

Brand: Thermo Scientific Acros 180621000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 546-67-8 | C8H12O8Pb | 443.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-67-8, 64-19-7
Molecular Formula C8H12O8Pb
Molecular Weight (g/mol) 443.36
InChI Key NVTAREBLATURGT-UHFFFAOYSA-N
Synonym lead iv acetate
PubChem CID 50931538
IUPAC Name acetic acid;lead
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]

Specifications

Melting Point 175.0°C to 180.0°C
Color Orange-Pink to White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 94% min. (Iodometry)
Linear Formula (CH3CO2)4Pb
Fieser 01,537; 02,234; 03,168; 04,278; 05,365; 06,313; 08,269; 09,265; 10,228; 12,270; 13,62; 14,188; 16,193
Merck Index 15,5478
Solubility Information Solubility in water: decomposes in water forming pb02 and acetic acid. Other solubilities: soluble in hot glacial acetic acid, soluble in benzene, chloroform, tetrachloroethane,, toluene, acetonitrile, nitrobenzene and, dichloromethane
Formula Weight 443.36
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Lead(IV) acetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(IV) acetate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.