missing translation for 'onlineSavingsMsg'
Learn More

Lead in Isooctane standard solution, Specpure™, 37μg/g(0.1g/gal), Thermo Scientific Chemicals

Product Code. 15476895
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 540-84-1
Quantity 100 g
Molecular Formula C8H18
MDL Number MFCD01094200
UN Number UN1262
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.232
PubChem CID 10907
ChEBI CHEBI:62805
Grade Specpure™
Odor Nearly odorless
Physical Form Liquid
Chemical Name or Material Lead in Isooctane standard solution, 37μg/g(0.1g/gal)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead in Isooctane standard solution, Specpure, 37µg/g (0.100g/gal)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Isooctane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'