missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead in Isooctane standard solution, Specpure™, 37.0μg/g(0.100g/gal)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 540-84-1 |
| Odor | Nearly odorless |
| Physical Form | Liquid |
| Molecular Formula | C8H18 |
| MDL Number | MFCD01094200 |
| UN Number | UN1262 |
| Color | Colorless |
| InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
| SMILES | CC(C)CC(C)(C)C |
| IUPAC Name | 2,2,4-trimethylpentane |
| Show More |
Safety and Handling
Product Identifier
- Lead in Isooctane standard solution, Specpure, 37.0µg/g (0.100g/gal)
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contain: Isooctane
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?