missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) zirconium oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 15446345
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446345 250 g 250g
11385047 10 g 10g
11375047 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15446345 Supplier Thermo Scientific Alfa Aesar Supplier No. 040754.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-01-4
Molecular Formula O3PbZr
Molecular Weight (g/mol) 346.421
MDL Number MFCD00016275
InChI Key RSCSNKLWUCUPDE-UHFFFAOYSA-N
Synonym lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate
PubChem CID 16212542
IUPAC Name dioxido(oxo)zirconium;lead(2+)
SMILES [O-][Zr](=O)[O-].[Pb+2]

Specifications

Physical Form Powder
Quantity 250 g
UN Number UN3077
Formula Weight 346.42
Percent Purity 99.997%
Grade Puratronic
Odor Odorless
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) zirconium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.