missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is employed in pyrolytic deposition of fluoride glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 147°C to 153°C |
Color | White |
Odor | Odorless |
Linear Formula | Pb(CF3COO)2·0.5H2O |
Quantity | 10 g |
UN Number | UN2291 |
Solubility Information | Soluble in water,trifluoroacetic acid,tetrahydrofuran,acetone. |
Formula Weight | 442.23(433.22 Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Lead(II) trifluoroacetate hemihydrate |
Safety and Handling
Product Identifier
- Lead(II) trifluoroacetate hemihydrate
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'