missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals

Product Code. 11353377
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353377 50 g 50g
11343377 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353377 Supplier Thermo Scientific Alfa Aesar Supplier No. 017553.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is employed in pyrolytic deposition of fluoride glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 4146-73-0
Molecular Formula C4F6O4Pb
Molecular Weight (g/mol) 433.23
MDL Number MFCD00061594
InChI Key XVULNTYAIWEMSW-UHFFFAOYSA-L
Synonym lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii
PubChem CID 71311182
IUPAC Name lead(2+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2]

Especificaciones

Melting Point 147°C to 153°C
Color White
Odor Odorless
Linear Formula Pb(CF3COO)2·0.5H2O
Quantity 50 g
UN Number UN2291
Formula Weight 442.23 (433.22 Anhydrous)
Physical Form Powder
Chemical Name or Material Lead(II) trifluoroacetate hemihydrate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.