missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) titanium oxide, 99.5%

Product Code. 11387737
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387737 100 g 100g
11397737 500 g 500g
11307747 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387737 Supplier Thermo Scientific Alfa Aesar Supplier No. 014139.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12060-00-3 | O3PbTi | 303.064 g/mol

Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-00-3
Molecular Formula O3PbTi
Molecular Weight (g/mol) 303.064
MDL Number MFCD00011159
InChI Key NKZSPGSOXYXWQA-UHFFFAOYSA-N
Synonym lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
PubChem CID 16211560
IUPAC Name dioxido(oxo)titanium;lead(2+)
SMILES [O-][Ti](=O)[O-].[Pb+2]

Specifications

Color Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 303.09
Percent Purity 99.5%
Odor Odorless
Density 7.520 g/mL
Chemical Name or Material Lead(II) titanium oxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) titanium oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.