missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide, 99% (metals basis), 50% w/v in isopropanol, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 25 mL |
UN Number | UN1993 |
Sensitivity | Moisture sensitive |
Formula Weight | 777.4 |
Percent Purity | 99% |
Physical Form | Liquid |
Assay | (metals basis) |
Chemical Name or Material | Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide |
Safety and Handling
Product Identifier
- Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide 50% w/v in isopropanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H360-May damage fertility or the unborn child.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : 2-Propanol, Lead(II)titanium(IV) 2-ethylhexanoate tetraisopropoxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'