missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) tartrate, 99%, Thermo Scientific™

Product Code. 11469543
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469543 25 g 25g
11479543 100 g 100g
11489543 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469543 Supplier Thermo Scientific Alfa Aesar Supplier No. A12388.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a raw material in organic chemistry for the synthesis of other chiral molecules. The acid has served in the farming and metal industries as a chelating agent for complexing micronutrients.

Solubility
Soluble in dilute nitric acid, alkali, acid ammonium hydrogen oxidation and tartaric acid ammonium solution, insoluble in water and alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815-84-9
Molecular Formula C4H4O6Pb
Molecular Weight (g/mol) 355.27
MDL Number MFCD00043282
InChI Key AYPZCTCULRIASE-ZVGUSBNCSA-L
Synonym unii-x5kmm6t3a6, lead tartrate, x5kmm6t3a6, lead tartrate pbc4h4o6, lead ii tartrate 1:1, lead tartrate van, butanedioic acid, 2,3-dihydroxy-2r,3r-, lead 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-r-r*,r*-, lead 2+ salt 1:1, +-l-tartaric acid lead ii salt, tartaric acid, lead 2+ salt 1:1 8ci
PubChem CID 101283265
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioate;lead(2+)
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.[Pb+2]

Specifications

Density 2.53
Quantity 25 g
UN Number UN2291
Solubility Information Soluble in dilute nitric acid,alkali,acid ammonium hydrogen oxidation and tartaric acid ammonium solution,insoluble in water and alcohol.
Formula Weight 355.27
Percent Purity 99%
Chemical Name or Material Lead (II) tartrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) tartrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.