missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) tantalum oxide, 99.9% (metals basis), Thermo Scientific™

Product Code. 11398108
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398108 100 g 100g
11388108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398108 Supplier Thermo Scientific Alfa Aesar Supplier No. 039348.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12065-68-8
Molecular Formula O6PbTa2
Molecular Weight (g/mol) 665.09
MDL Number MFCD00054256
InChI Key KNSIKTVGKTWOGA-UHFFFAOYSA-N
Synonym lead ii tantalum oxide, lead tantalate, lead ii tantalate
PubChem CID 16685095
IUPAC Name (dioxotantaliooxy-$l^{2}-plumbanyl)oxy-dioxotantalum
SMILES O=[Ta](=O)O[Pb]O[Ta](=O)=O

Specifications

Melting Point >350°C
Physical Form Powder
Quantity 100 g
UN Number UN2291
Formula Weight 665.09
Percent Purity 99.9%
Density 7.9 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) tantalum oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.