missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfide, Pb 82% min.

Product Code. p-7080651
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11413993

Brand: Thermo Scientific Alfa Aesar A10268.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-87-0 | PbS | 239.30 g/mol

Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
SMILES [S--].[Pb++]

Specifications

Melting Point 1114°C
Quantity 500 g
UN Number UN3077
Merck Index 14,5421
Solubility Information Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide.
IUPAC Name λ²-lead(2+) sulfanediide
Formula Weight 239.25
Concentration or Composition (by Analyte or Components) Pb 82% min.
Odor Odorless
Refractive Index 3.921
Density 7.5
Chemical Name or Material Lead (II) sulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) sulphide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Lead(II) sulphide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.