missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11400061
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400061 500 g 500g
11490051 100 g 100g
11480051 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400061 Supplier Thermo Scientific Alfa Aesar Supplier No. 044784.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Pigments, Galvanic Batteries, Lithography

Lead(II) sulfate is used in pigments, galvanic batteries, lithography, weighting fabrics. It is also used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-14-2
Molecular Formula O4PbS
Molecular Weight (g/mol) 303.256
MDL Number MFCD00011166
InChI Key KEQXNNJHMWSZHK-UHFFFAOYSA-L
Synonym lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
PubChem CID 24008
IUPAC Name lead(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 1170°C
Color White
Physical Form Lump/Powder
Quantity 500 g
Solubility Information Partially soluble in water,dilute HCl and HNO3. Soluble in sodium hydroxide,ammonium acetate or tartrate,concentrated hydriodic acid. Insoluble in alcohol
Formula Weight 303.25
Grade Reagent
Odor Odorless
Appearance White powder/lump
Density 6.2
Chemical Name or Material Lead(II) sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.