missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfate, 99%

Product Code. 10648614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10648614 500 g Plastic bottle 500g
10686923 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10648614 Supplier Thermo Scientific Acros Supplier No. 387275000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-14-2 | O4PbS | 303.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-14-2
Molecular Formula O4PbS
Molecular Weight (g/mol) 303.25
MDL Number MFCD00011166
InChI Key KEQXNNJHMWSZHK-UHFFFAOYSA-L
Synonym lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
PubChem CID 24008
IUPAC Name lead(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 1170.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Merck Index 15, 5475
Solubility Information Solubility in water: slightly soluble
Formula Weight 303.25
Percent Purity 99%
Identification Confirmed
Packaging Plastic bottle
Chemical Name or Material Lead(II) sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.