missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) selenide, 99.999+% (metals basis)

Product Code. 15452067
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452067 100 g 100g
15442067 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452067 Supplier Thermo Scientific Alfa Aesar Supplier No. 044916.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12069-00-0 | PbSe | 286.20 g/mol

Lead(II) selenide is used in the production of infrared detectors for thermal imaging. It is also used as quantum dots in nanocrystal solar cell. It is an important material and finds applications in photographic plates, and photo resistors and optoelectronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-00-0
Molecular Formula PbSe
Molecular Weight (g/mol) 286.20
MDL Number MFCD00016273
InChI Key GGYFMLJDMAMTAB-UHFFFAOYSA-N
Synonym lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g
PubChem CID 61550
IUPAC Name selanylidenelead
SMILES [Se]=[Pb]

Specifications

Color Gray
Physical Form Fused Lump
Quantity 100 g
Assay Percent Range >99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 286.15
Chemical Name or Material Lead(II) selenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.