missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Oxide, TRC

Product Code. 30479154
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479154 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479154 Supplier Toronto Research Chemicals Supplier No. TRCL32002510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lead(II) oxide
CAS 1317-36-8
Quantity 10 g
Molecular Formula PbO
Synonym Lead oxide (PbO), C.I. 77577, C.I. Pigment Yellow 46, G 2, Lead monooxide, Lead monoxide, Lead oxide, Lead oxide yellow, Lead protoxide, Lead(2+) oxide, Lead(II) oxide, Litharge, Litharge S, Litharge Yellow L 28, Pigment Yellow 46, Plumbous oxide, Yellow lead ocher
InChI Formula InChI=1S/O.Pb
SMILES O=[Pb]
IUPAC Name oxolead
Molecular Weight (g/mol) 223
Formula Weight 223.9716
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.