missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, Puratronic™, 99.9995% (metals basis)

Product Code. 11326807
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326807 500 g 500g
11306807 25 g 25g
11316807 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326807 Supplier Thermo Scientific Alfa Aesar Supplier No. 010728.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1317-36-8 | OPb | 223.20 g/mol

It is used in the manufacture of glass, ceramics and computer components. Addition of lead oxide increases the refractive index, electrical resistivity, Curie temperature, and the ability of the glass to absorb X-rays. It is also added to decrease the viscosity of glass. It is employed in the vulcanization of rubber, lead acid batteries, and in the production of certain pigments and paints. In cathode ray tube glass lead oxide is used to block X-ray emission. It acts as an efficient reagent in oxidation reactions as well as condensation reactions of hydroxy ketones with diamines to form quinoxaline derivatives with high yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Melting Point 886°C
Color Yellow
Physical Form Powder
Quantity 500 g
Linear Formula PbO
UN Number UN3077
Merck Index 14,5413
IUPAC Name plumbanone
Formula Weight 223.19
Percent Purity 99.9995%
Grade Puratronic
Odor Odorless
Density 9.53g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.