missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, meets analytical spec. of DAC, BPC, 99 to 100.5% (complexometric), yellow, Honeywell Fluka™

Product Code. p-7122042 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Fiberboard Box
Poly bottle
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15651060 1 kg Poly bottle 1kg
15661060 25 kg Fiberboard Box 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15651060 Supplier Honeywell-Fluka Supplier No. 115261KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meets analytical specification of DAC, BPC, 99-100.5% (complexometric), yellow

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name oxolead
SMILES O=[Pb]

Specifications

Color Yellow
Quantity 1 kg
Linear Formula PbO
UN Number 2291
Formula Weight 223.2g/mol
Percent Purity 99 to 100.5% (complexometric)
Packaging Poly bottle
Vapor Pressure 10mmHg (0°C)
Chemical Name or Material Lead(Ii) Oxide, Meets analytical specification of DAC, BPC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.