missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, 99.99% (metals basis)

Product Code. 11347807
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347807 1 kg 1kg
11357807 50 g 50g
11337807 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347807 Supplier Thermo Scientific Alfa Aesar Supplier No. 014240.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-36-8 | OPb | 223.20 g/mol

Lead(II) oxide is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. It is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Melting Point 886°C
Color Yellow
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in concentrated alkali,hydrochloric acid,and ammonium chloride. Insoluble in water,dilute alkali and alcohol.
IUPAC Name plumbanone
Formula Weight 223.19
Odor Odorless
Appearance Yellow powder
Density 9.53
Chemical Name or Material Lead(II) oxide
Show More Show Less
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity on or via lactation
Hazard Statement
  • H351-Suspected of causing cancer.
  • H362-May cause harm to breast-fed children.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.