missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Nitrate, TRC

Product Code. 30483469
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
Unit Size:
10g
250g
25g
Product Code. Quantity unitSize
30483469 250 g 250g
30482651 25 g 25g
30479251 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 30483469

Brand: Toronto Research Chemicals TRCL178800250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8
Formula Weight 331.9523
InChI Formula InChI=1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2
IUPAC Name lead(2+);dinitrate
Molecular Formula 2 N O3 . Pb
Molecular Weight (g/mol) 331.21
SMILES [Pb+2].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
Synonym Lead(II) Nitrate
Chemical Name or Material Lead nitrate

Specifications

Quantity 250 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.