missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) nitrate, Puriss., ≥99%, Solstice

Product Code. 15641060 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
50 kg
Unit Size:
1kg
500g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15641060 50 kg 50kg
15631060 500 g 500g
15621060 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15641060 Supplier Honeywell-Fluka Supplier No. 1152050KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187

Specifications

Melting Point 470°C (decomposition)(literature)
CAS Max % 100.0000%
Quantity 50 kg
Linear Formula Pb(NO3)2
UN Number UN1469
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
IUPAC Name λ²-lead(2+) dinitrate
Formula Weight 331.21
Percent Purity >99%
Grade puriss.
Packaging Fiberboard Box
Density 4.49 g/cm3
Chemical Name or Material Lead(II) nitrate, Puriss., ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.