missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Nitrate, ACS Reagent, Solstice

Product Code. 15632270
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15632270 500 g 500g
15612270 100 g 100g
15622270 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15632270 Supplier Honeywell-Fluka Supplier No. 228621500G
  • ONLINE EXCLUSIVE PRICE
    £67.00
    £61.62 / 500g
    Save £5.38
    8% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name lead(2+);dinitrate

Spezifikation

Physical Form Solid
Quantity 500 g
Linear Formula Pb(NO3)2
UN Number 1469
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
Formula Weight 331.21g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Lead(Ii) Nitrate
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Lead(II) nitrate
Signalwort
  • Danger
Gefahrenkategorie
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Reproductive toxicity Category 1A
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Gefahrenhinweis
  • H272-May intensify fire; oxidiser.
  • H302+H332-Harmful if swallowed or if inhaled
  • H318-Causes serious eye damage.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.