missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
78.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
Description
As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photographyEmployed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 470°C (decomposition) |
Color | White |
Physical Form | Crystals |
Quantity | 2 kg |
Assay Percent Range | 99.0% min. |
Solubility Information | Freely soluble in water. Soluble in absolute alcohol,methanol. Insoluble in concentrated HNO3 |
IUPAC Name | λ²-lead(2+) dinitrate |
Formula Weight | 331.2 |
Grade | ACS |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'