missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) metavanadate, 99.9% (metals basis), Thermo Scientific™

Product Code. 11388148
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388148 250 g 250g
11378148 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388148 Supplier Thermo Scientific Alfa Aesar Supplier No. 039378.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As pigment, in manufacturing of vanadium compounds

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-79-3
Molecular Formula O6PbV2
Molecular Weight (g/mol) 405.10
MDL Number MFCD00016271
InChI Key OGRLITDAVSILTM-UHFFFAOYSA-N
Synonym lead vanadate, unii-99565ywk3f, lead ii metavanadate, lead vanadate v, lead divanadium hexaoxide, lead vanadium oxide pbv2o6, 2o3v.pb, lead 2+ ; oxido dioxo vanadium, $l^ 2-lead 2+ ion divanadiumoylolate
PubChem CID 61471
IUPAC Name lead(2+);oxido(dioxo)vanadium
SMILES [Pb++].[O-][V](=O)=O.[O-][V](=O)=O

Specifications

Physical Form Powder
Quantity 250 g
UN Number UN2291
Merck Index 14,5427
Solubility Information Insoluble in water,decomposes in HNO3
Formula Weight 405.07
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lead(II) metavanadate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.