missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) metaborate monohydrate, Thermo Scientific Chemicals

Product Code. 11328108
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328108 25 g 25g
11338108 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328108 Supplier Thermo Scientific Alfa Aesar Supplier No. 039339.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lead(II) metaborate monohydrate acts as a starting material used in the preparation of lead(II) borates such as Pb2B3O5.5 (OH)2 and [Pb3(B3O7)](NO3) by changing the pH values of the reaction systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10214-39-8
MDL Number MFCD00049504
Synonym Lead(II) borate

Specifications

Melting Point 160°C (in -H2O)
Physical Form Powder
Quantity 25 g
Linear Formula PbB2O4·H2O
UN Number UN2291
Solubility Information Insoluble in water and alkali; Soluble in acids.
Formula Weight 310.81 (292.81 Anhydrous)
Odor Odorless
Density 5.598 g/mL
Chemical Name or Material Lead(II) metaborate monohydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) metaborate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.